Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNMT Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB102912 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB102912 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB102912 Supplier Novus Biologicals Supplier No. NBP297644AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNMT Polyclonal specifically detects RNMT in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RNMT
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias DKFZp686H1252, EC 2.1.1.56, hcm1p, hCMT1, hCMT1c, hMet, KIAA0398, mRNA (guanine-7-)methyltransferase, mRNA (guanine-N(7)-)-methyltransferase, mRNA cap guanine-N7 methyltransferase, mRNA cap methyltransferase, RG7MT1MET, RNA (guanine-7-) methyltransferase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RNMT fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8731
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.