Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7205766 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8976 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8976 Supplier Proteintech Group Inc Supplier No. 137431AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNMT belongs to the mRNA cap methyltransferase family. RNMT is a mRNA-capping methyltransferase that methylates the N7 position of the added guanosine to the 5′-cap structure of mRNAs. It binds RNA containing 5′-terminal GpppC.

Specifications

Antigen RNMT
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNMT
Gene Accession No. O43148, Q5U2U7, Q9D0L8
Gene Alias hcm1p, hCMT1, hCMT1c, hMet, KIAA0398, MET, mRNA cap methyltransferase, RG7MT1, RNMT
Gene Symbols Rnmt
Host Species Rabbit
Immunogen RNMT Fusion Protein Ag4682
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291534, 67897, 8731
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.