Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNPC3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157138
Description
RNPC3 Polyclonal specifically detects RNPC3 in Human samples. It is validated for Western Blot.Specifications
RNPC3 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ20008, FLJ25070, KIAA183, KIAA1839, RBM40U11/U12 snRNP 65 kDa protein, RNA recognition protein, RNA-binding motif protein 40, RNA-binding protein 40, RNA-binding region (RNP1, RRM) containing 3, RNA-binding region-containing protein 3, RNP, U11/U12 small nuclear ribonucleoprotein 65 kDa protein, U11/U12 snRNP 65K, U11/U12-65K | |
Rabbit | |
Affinity purified | |
RUO | |
55599 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q96LT9 | |
RNPC3 | |
Synthetic peptides corresponding to RNPC3(RNA-binding region (RNP1, RRM) containing 3) The peptide sequence was selected from the middle region of RNPC3. Peptide sequence LHAPLPPTSPQPPEEPPLPDEDEELSSEESEYESTDDEDRQRMNKLMELA. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Bovine: 92%; Canine: 92%; Pig: 92%; Mouse: 84%; Rat: 84%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit, Yeast | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction