Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNPEPL1 Antibody (4A9), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB259371700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259371700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259371700 Supplier Novus Biologicals Supplier No. NBP259371AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNPEPL1 Monoclonal specifically detects RNPEPL1 in Non-species specific samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen RNPEPL1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 4A9
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1
Gene Symbols RNPEPL1
Host Species Mouse
Immunogen Synthetic N-terminal arginylated peptide conjugated to KLH
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 57140
Test Specificity Specific for N-terminal arginine next to glutamic acid. Does not detect N-terminal arginine next to aspartic acid or internal arginine residues.
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.