Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNPEPL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | RNPEPL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RNPEPL1 Polyclonal specifically detects RNPEPL1 in Human samples. It is validated for Western Blot.Specifications
RNPEPL1 | |
Polyclonal | |
Rabbit | |
Q9HAU8 | |
57140 | |
Synthetic peptides corresponding to RNPEPL1(arginyl aminopeptidase (aminopeptidase B)-like 1) The peptide sequence was selected from the N terminal of RNPEPL1. Peptide sequence LKPADIGPRSRVWAEPCLLPTATSKLSGAVEQWLSAAERLYGPYMWGRYD. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
argininyl aminopeptidase-like 1, arginyl aminopeptidase (aminopeptidase B)-like 1, arginyl aminopeptidase-like 1, EC 3.4.11.-, FLJ10806, FLJ26675, MGC99544, RNPEP-like 1 | |
RNPEPL1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title