Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ro 08-2750
GREENER_CHOICE

Catalog No. 227210
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-721-0 10 mg
22-721 1 mg
22-725-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-721-0 Supplier Tocris Bioscience™ Supplier No. 2272/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibits NGF binding to p75NTR and TrkA

Non-peptide inhibitor of NGF that binds the NGF dimer (KD ∽ 1 μM) possibly causing a conformational change. Selectively inhibits binding of NGF to p75NTR at submicromolar concentrations, and to both p75NTR and TrkA at concentrations > 5 μM. Prevents NGF-induced apoptosis in SK-N-MC cells. Suppresses cancer cell proliferation via regulation of Hippo signaling aand YAP phosphorylation.

Chemical Identifiers

CAS 37854-59-4
Molecular Formula C13H10N4O3

Specifications

Quantity 10 mg
Formula Weight 270.24
Purity >96%
Molecular Weight (g/mol) 270.248
Chemical Name or Material Ro 08-2750
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.