Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ro 31-8220 mesylate
GREENER_CHOICESDP

Catalog No. 200210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-021-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 20-021-0 Supplier Tocris Bioscience™ Supplier No. 2002/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Protein kinase inhibitor

Protein kinase C inhibitor, with activity at other protein kinases (IC50 values are 33, 3, 8, 15 and 38 nM for PKCα, MAPKAP-K1b, MSK1, GSK3β and S6K1 respectively). Activates JNK and glycogen synthase, and inhibits MAPK and ERK2, in rat adipocytes and L6 myotubes. Also inhibits voltage-dependent Na+ channels in the micromolar range. Induces protein-protein interaction between mutant SMAD4R361H and SMAD3, leading to reactivation of dormant SMAD4R361H-mediated transcriptional activity and restoration of TGF-β-induced tumor suppression activity in SMAD4 mutant cancer cells.

Chemical Identifiers

CAS 138489-18-6
Molecular Formula C26H27N5O5S2

Specifications

Quantity 10 mg
Formula Weight 553.65
Purity >97%
Molecular Weight (g/mol) 553.652
Chemical Name or Material Ro 31-8220 mesylate
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.