Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ro 51, Tocris Bioscience™

Catalog No. 439150
Change view
Click to view available options
Quantity:
10 mg
50 mg
Catalog No. Quantity
43-915-0 50 mg
43-911-0 10 mg
2 options
This item is not returnable. View return policy

Catalog No. 43-915-0

Supplier: R&D Systems 4391/50

This item is not returnable. View return policy

Potent P2X3, P2X2/3 antagonist

Potent, dual P2X3 and P2X2/3 antagonist (IC50 values are 2 and 5 nM for rP2X3 and hP2X2/3 respectively). Selective for P2X3 and P2X2/3 over other P2X receptors (IC50 values are > 10 μM for P2X1, P2X2, P2X4, P2X5, and P2X7). Cell permeable.

Chemical Identifiers

CAS 1050670-85-3
Molecular Formula C17H23IN4O4
Molecular Weight (g/mol) 474.299
InChI Key PAYROHWFGZADBR-UHFFFAOYSA-N
Synonym 2-4-amino-5-5-iodo-2-isopropyl-4-methoxyphenoxy pyrimidin-2-yl amino propane-1,3-diol, compound 128 pmid: 26686393, 2-4-amino-5-5-iodo-2-isopropyl-4-methoxyphenoxy pyrimidin-2-ylamino propane-1,3-diol, 2-4-amino-5-5-iodo-2-isopropyl-4-methoxy-phenoxy-pyrimidin-2-ylamino-propane-1,3-diol, 2-4-amino-5-5-iodo-4-methoxy-2-propan-2-ylphenoxy pyrimidin-2-yl amino propane-1,3-diol, 2-4-amino-5-5-iodo-4-methoxy-2-1-methylethyl phenoxy-2,4-pyrimidinyl amino-1,3-propanediol
PubChem CID 24883277
IUPAC Name 2-[[4-amino-5-(5-iodo-4-methoxy-2-propan-2-ylphenoxy)pyrimidin-2-yl]amino]propane-1,3-diol
SMILES CC(C)C1=CC(=C(C=C1OC2=CN=C(N=C2N)NC(CO)CO)I)OC

Specifications

Quantity 50 mg
Formula Weight 474.29
Percent Purity >98%
Chemical Name or Material Ro 51
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.