Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ RO 5263397 hydrochloride

Click to view available options
Quantity:
5 mg
25 mg
Description
RO 5263397 hydrochloride is a potent trace amine 1 (TA1) receptor agonist (EC50 values are 0.12, 35 and 17-85 nM for mouse, rat and human receptors, respectively). Increases wakefulness and reduces REM and NREM sleep duration in wild type mice. Inhibits spontaneous locomotor activity in dopamine transport (DAT) knockout mice.
Specifications
Specifications
Chemical Name or Material | (4S)-4-(3-Fluoro-2-methylphenyl)-4,5-dihydro-2-oxazolamine hydrochloride |
Quantity | 25 mg |
Target | Trace Amine 1 (TA1) Receptor Agonists |
Molecular Formula | C10H11FN2O.HCl |
Formula Weight | 3131 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction