Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ro 64-5229
GREENER_CHOICE

Catalog No. 291310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-131-0 10 mg
29-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-131-0 Supplier Tocris Bioscience™ Supplier No. 2913/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective, non-competitive mGlu2 antagonist

Selective, non-competitive mGlu2 antagonist. Inhibits GTPγ35S binding to mGlu2-containing membranes (IC50 = 0.11 μM).

Chemical Identifiers

CAS 246852-46-0
Molecular Formula C17H19Cl2N3O

Specifications

Quantity 10 mg
Formula Weight 352.26
Purity >99%
Molecular Weight (g/mol) 352.259
Chemical Name or Material Ro 64-5229
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.