Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ro 6842262, >98%
GREENER_CHOICESDP

Catalog No. 591310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-131-0 10 mg
59-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-131-0 Supplier Tocris Bioscience™ Supplier No. 5913/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent LPA1 antagonist

Potent LPA1 antagonist (IC50 = 25 nM). Exhibits >1,200-fold selectivity for LPA1 over LPA3. Attenuates NHLF human lung fibroblast cell proliferation and contraction in vitro. Also reduces plasma histamine levels in mouse LPA-challenge model. Orally bioavailable.

Chemical Identifiers

CAS 1396006-71-5
Molecular Formula C28H26N4O4

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Formula Weight 482.53
Purity >98%
Molecular Weight (g/mol) 482.54
Chemical Name or Material 1-[4'-[4-Methyl-5-[[[(1R)-1-phenylethoxy]carbonylamino]-1H-1,2,3-triazol-1-yl][1,1'-biph
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.