Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROBO1 Antibody, Novus Biologicals™
SDP

Catalog No. NB6001254 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001254 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001254 Supplier Novus Biologicals Supplier No. NB6001254
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ROBO1 Polyclonal specifically detects ROBO1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen ROBO1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA 1:30000-1:60000, Immunohistochemistry 2 ug/ml-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q9Y6N7
Gene Alias DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila)
Gene Symbols ROBO1
Host Species Rabbit
Immunogen This affinity-purified ROBO1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an C-Terminal region near amino acids 1625-1650 of Human ROBO1. (Uniprot: Q2M1J3)
Molecular Weight of Antigen 175.89 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6091
Test Specificity This affinity purified antibody is directed against human ROBO1 protein. A BLAST analysis was used to suggest reactivity with this protein from human, mouse, rat and dog sources based on 100% homology for the immunogen sequence. Cross reactivity will occur with all isoforms of ROBO1. Cross reactivity with ROBO1 homologues from other sources has not been determined.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.