Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROBO2 Antibody (356001), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB348508 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348508 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348508 Supplier Novus Biologicals Supplier No. FAB3147MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROBO2 Monoclonal specifically detects ROBO2 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen ROBO2
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 356001
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Drosophila) homolog 2, roundabout, axon guidance receptor, homolog 2 (Drosophila)
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ROBO2 isoform 1 Ser22-Pro859 Accession # Q9HCK4.2
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6092
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.