Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROBO2 Antibody, Novus Biologicals™
SDP

Catalog No. NB387859 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387859 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387859 Supplier Novus Biologicals Supplier No. NBP323374
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ROBO2 Polyclonal antibody specifically detects ROBO2 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot

Specifications

Antigen ROBO2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias Drosophila) homolog 2, roundabout, axon guidance receptor, homolog 2 (Drosophila)
Host Species Rabbit
Immunogen Robo2 synthetic peptide (coupled to carrier protein) corresponds to amino acids in the C-terminal region of human Robo2. This sequence has significant homology to the same region in rat and mouse Robo2, and has low homology to other Robo family members. [UniProt# Q9HCK4]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6092
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.