Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROBO4 Antibody (265703), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB313445 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313445 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313445 Supplier Novus Biologicals Supplier No. FAB2454PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROBO4 Monoclonal specifically detects ROBO4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen ROBO4
Applications Flow Cytometry
Classification Monoclonal
Clone 265703
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias ECSM4, FLJ20798, Magic roundabout, MGC133352, MGC133353, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout (Drosophila)
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ROBO4, Gln28-Arg467, Accession # Q8WZ75
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Extracellular Matrix, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54538
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.