Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROBO4 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB339177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339177 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339177 Supplier Novus Biologicals Supplier No. NB11058780PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ROBO4 Polyclonal antibody specifically detects ROBO4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen ROBO4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ECSM4, FLJ20798, Magic roundabout, MGC133352, MGC133353, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout (Drosophila)
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human Robo4 (within residues 900-1007). [Swiss-Prot# Q8WZ75]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Extracellular Matrix, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54538
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.