Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human and Mouse. Others not known.
Epitope: aa 97-108
Immunogen: A peptide corresponding to amino acid residue 97-108 (CPLDNREWEFQKYGH) from C-terminus of the human ROC1
Molecular Weight: 13kDa
Positive Control: HeLa cells
Cellular Localization: Nuclear
Recommended for:
- Western Blotting
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | ROC1 Ab-1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A peptide corresponding to amino acid residue 97-108 (CPLDNREWEFQKYGH) from C-terminus of the human ROC1 |
| Purification Method | Purified with BSA and azide |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.