Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROC1 Ab-1 Rabbit Polyclonal Antibody, Epredia™

Catalog No. rb069p1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Purification Method:
Purified w/o BSA and azide
Purified with BSA and azide
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Purification Method
RB069P1 500 μL Purified with BSA and azide
RB069P0 100 μL Purified with BSA and azide
RB069P 1 mL Purified with BSA and azide
RB069P1ABX 500 μL Purified w/o BSA and azide
RB069PABX 1 mL Purified w/o BSA and azide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. RB069P1 Supplier Epredia Supplier No. RB069P1

Provide accurate, reproducible results with the Epredia™ ROC1 Ab-1, Rabbit Polyclonal Antibody.

ROC1 and ROC2 are the two highly conserved RING finger proteins that are homologous to APC11, a subunit of the anaphase-promoting complex(APC). ROC1 and ROC2 commonly interact with all cullins while APC11 specifically interacts with APC2, a cullin-related APC subunit. ROC1 and APC11 immunocomplexes can catalyze isopeptide ligations to form polyubiquitin chains in an E1- and E2-dependent manner. ROC1 is thought to play a unique role in the ubiquitination reaction by heterodimerizing with cullin1 to catalyze ubiquitin polymerization.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not known.

Epitope: aa 97-108

Immunogen: A peptide corresponding to amino acid residue 97-108 (CPLDNREWEFQKYGH) from C-terminus of the human ROC1

Molecular Weight: 13kDa

Positive Control: HeLa cells

Cellular Localization: Nuclear

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen ROC1 Ab-1
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen A peptide corresponding to amino acid residue 97-108 (CPLDNREWEFQKYGH) from C-terminus of the human ROC1
Purification Method Purified with BSA and azide
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.