Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Roche Diagnostics CoaguChek XS Pro System Catalog No.: 22045351
Only null left
Add to Cart

Roche Diagnostics CoaguChek XS Pro System

Catalog No. 22045351
Each
Click to view available options
Quantity:
Each
This item is not returnable. View return policy

Catalog No. 22-045-351

Supplier: Roche Diagnostics 05530199160

Only null left
Add to Cart
Only null left
Add to Cart
This item is not returnable. View return policy

For quantitative prothrombin time testing for the monitoring of warfarin therapy

Barcode scanning technology

  • Quickly capture patient and operator ID information
  • Helps enhance patient safety and save time by eliminating the need to manually enter ID information
Accurate and flexible technology
  • Accurate results in 1 minute with 97% correlation to the lab method using the Dade Sysmex 560 Analyzer and Dade Innovin
  • Performs patient results and onboard controls in a single test chamber on the test strip
  • Neutralizes therapeutic levels of heparin and LMWH
  • INR corrected for hematocrit within specified range
  • Streamlined data management
  • Stores 1000 patient and 500 optional liquid QC results

  • Operator and QC lockout capabilities
  • Data storage for up to 60 code chips
  • Easy and convenient to use
  • Small sample size (8μL) from a finger stick
  • Top or side strip dosing, with outside-meter blood application

Connectivity between the CoaguChek XS Plus meter and the RALS-Plus data management software is enabled via the Handheld Base Unit

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Capillary Whole Blood
Type CoaguChek™ XS Pro Prothrombin Testing System
For Use With (Application) Quantitative Prothrombin time testing for the monitoring of warfarin therapy
Includes Barcode scanning, accurate and flexible technology
Quantity Each
Sample Size 8 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Roche Diagnostics CoaguChek XS Pro System >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.