Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK1/2 Antibody (HL1632) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294975 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294975 Supplier Novus Biologicals Supplier No. NBP325642
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ROCK1/2 Monoclonal antibody specifically detects ROCK1/2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ROCK1/2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HL1632
Conjugate Unconjugated
Dilution Western Blot 1:500-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias p160ROCK, p164 ROCK-2, Rho associated coiled-coil containing protein kinase 1, Rho associated coiled-coil containing protein kinase 2, ROCK-I, ROCK-II
Host Species Rabbit
Immunogen Recombinant fragment of human ROCK1 .
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6093
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.