Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK1 Antibody (12M03), Novus Biologicals™
SDP

Catalog No. NB018601 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018601 0.025 mg
NBP146006 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018601 Supplier Novus Biologicals Supplier No. NBP1460060.025MG

Mouse Monoclonal Antibody

ROCK1 Monoclonal specifically detects ROCK1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen cleaved ROCK1
Applications Western Blot
Classification Monoclonal
Clone 12M03
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.2), 1% BSA, 50% Glycerol with 0.02% Thimerosal
Gene Accession No. Q13464
Gene Alias EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1
Gene Symbols ROCK1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the caspase-3 cleavage site.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6093
Test Specificity The antibody has been selected for its ability to recognize the cleaved ROCK-1 in Western blotting. It does not recognize the full length protein.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.