Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK1 Antibody (154C1465), DyLight 550 - (active/cleaved), Novus Biologicals™
SDP

Catalog No. NB10056596R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056596R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056596R Supplier Novus Biologicals Supplier No. NB10056596R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

cleaved ROCK1 Monoclonal specifically detects cleaved ROCK1 in Canine, Human, Mouse, Primate, Rabbit, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen cleaved ROCK1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 154C1465
Conjugate DyLight 550
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Accession No. O60259
Gene Alias EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1
Gene Symbols ROCK1
Host Species Mouse
Immunogen This antibody was generated by immunizing mice with a synthetic peptide sequence corresponding to amino acids 1114-1126 GNLPESRIEGWLS of human ROCK-1. This sequence is 100% conserved between human, chimpanzee, mouse, rat, dog, and rabbit. This immunogen peptide sequence is adjacent to the caspase-3 cleavage site of ROCK-1.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6093
Test Specificity This mAb recognizes the cleaved C-terminus of ROCK-1. It does not recognize the full-length protein. Since ROCK-1 is cleaved by caspase-3 during apoptosis, this may be used for detection of apoptosis.
Target Species Canine, Human, Rat, Mouse, Primate, Rabbit, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.