Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK1 Antibody (154C1465), Janelia Fluor™ 646 - (active/cleaved), Novus Biologicals™
SDP

Catalog No. N233238J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N233238J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N233238J646 Supplier Novus Biologicals Supplier No. NBP233238JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROCK1 Monoclonal specifically detects ROCK1 in Human, Mouse, Rat, Canine, Primate, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen ROCK1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 154C1465
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1
Gene Symbols ROCK1
Host Species Mouse
Immunogen This antibody was generated by immunizing mice with a synthetic peptide sequence corresponding to amino acids 1114-1126 GNLPESRIEGWLS of human ROCK-1. This sequence is 100% conserved between human, chimpanzee, mouse, rat, dog, and rabbit. This immunogen peptide sequence is adjacent to the caspase-3 cleavage site of ROCK-1.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Breast Cancer, Cancer, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6093.0
Test Specificity This mAb recognizes the cleaved C-terminus of ROCK-1. It does not recognize the full-length protein. Since ROCK-1 is cleaved by caspase-3 during apoptosis, this may be used for detection of apoptosis.
Target Species Human, Mouse, Rat, Canine, Primate, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.