Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK2 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB196910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB196910 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB196910 Supplier Novus Biologicals Supplier No. NBP176570C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ROCK2 Polyclonal antibody specifically detects ROCK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen ROCK2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias coiled-coil-containing protein kinase 2, EC 2.7.11, EC 2.7.11.1, KIAA0619, p164 ROCK-2, Rho kinase 2, rho-associated protein kinase 2, Rho-associated, coiled-coil containing protein kinase 2
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the center of human ROCK2. The immunogen is located within amino acids 920 - 970 of ROCK2. Amino Acid Squence: DLEKEKIMKELEIKE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 9475
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.