Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | ROD1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18045320
![]() |
Novus Biologicals
NBP18045320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180453
![]() |
Novus Biologicals
NBP180453 |
100 μL |
Each for $487.50
|
|
|||||
Description
ROD1 Polyclonal specifically detects ROD1 in Mouse samples. It is validated for Western Blot.Specifications
ROD1 | |
Polyclonal | |
Rabbit | |
NP_659153 | |
9991 | |
Synthetic peptide directed towards the N terminal of human Rod1. Peptide sequence YYTPVTPHLRSQPVYIQYSNHRELKTDNLPNQARAQAALQAVSAVQSGNL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DKFZp781I1117, fission yeast differentiation regulator, PTBP3, regulator of differentiation (in S. pombe) 1, regulator of differentiation 1, Rod1, ROD1 regulator of differentiation 1 (S. pombe) | |
PTBP3 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title