Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROPN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549502
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549502 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549502 Supplier Thermo Scientific Supplier No. PA549502
Only null left
Add to Cart
Add to Cart

ROPN1 Polyclonal Antibody

ROPN1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ROPN1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ROPN1
Gene Accession No. Q9HAT0
Gene Alias Cancer/testis antigen 91; CT91; ODF6; outer dense fiber of sperm tails 6; rhophilin associated tail protein 1; rhophilin-associated protein 1A; RHPNAP1; ROPN1; ROPN1A; ropporin; ropporin, rhophilin associated protein 1; ropporin-1A; testis secretory sperm-binding protein Li 239w
Gene Symbols ROPN1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 91-121 amino acids from the Central region of human ROPN1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54763
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.