Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roquin Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB190014 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB190014 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB190014 Supplier Novus Biologicals Supplier No. NBP281891AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Roquin Polyclonal antibody specifically detects Roquin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Roquin
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias KIAA2025RP5-1198E17.5, RING finger and C3H zinc finger protein 1, ring finger and CCCH-type domains 1, RING finger protein 198, RNF198ring finger and CCCH-type zinc finger domains 1, roquin
Host Species Rabbit
Immunogen Rabbit polyclonal Roquin antibody was raised against an 18 amino acid peptide near the carboxy terminus of human Roquin. The immunogen is located within amino acids 1040 - 1090 of Roquin. Amino Acid Squence: KLNTSQQAENGQPEPQNK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 149041
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.