Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR alpha/NR1F1 Antibody (784652), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB302514 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302514 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302514 Supplier Novus Biologicals Supplier No. FAB8924T

Mouse Monoclonal Antibody

ROR alpha/NR1F1 Monoclonal antibody specifically detects ROR alpha/NR1F1 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen ROR alpha/NR1F1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 784652
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias DKFZp686M2414, MGC119326, NR1F1MGC119329, nuclear receptor ROR-alpha, Nuclear receptor RZR-alpha, Nuclear receptor subfamily 1 group F member 1, RAR-related orphan receptor A, RAR-related orphan receptor alpha, retinoic acid receptor-related orphan receptor alpha, retinoid-related orphan receptor alpha, Retinoid-related orphan receptor-alpha, ROR1, ROR2, ROR3, RZR-ALPHA, RZRAROR-alpha, transcription factor RZR-alpha
Host Species Mouse
Immunogen E. coli-derived recombinant human ROR alpha/NR1F1, Arg47-His163, Accession # P35398
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6095
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.