Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR beta Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB045497 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045497 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045497 Supplier Novus Biologicals Supplier No. NBP297604DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ROR beta Polyclonal specifically detects ROR beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ROR beta
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias NR1F2nuclear receptor ROR-beta, Nuclear receptor RZR-beta, Nuclear receptor subfamily 1 group F member 2, RAR-related orphan receptor B, RAR-related orphan receptor beta, retinoic acid-binding receptor beta, retinoid-related orphan receptor beta, Retinoid-related orphan receptor-beta, ROR-BETA, RZRBbA133M9.1, RZR-BETA
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ROR beta fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 6096
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.