Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR gamma/RORC/NR1F3 Antibody (4G419), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NBP224451 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224451 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224451 Supplier Novus Biologicals Supplier No. NBP224451
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROR gamma/RORC/NR1F3 Monoclonal specifically detects ROR gamma/RORC/NR1F3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen ROR gamma/RORC/NR1F3
Applications Flow Cytometry
Classification Monoclonal
Clone 4G419
Conjugate APC
Dilution Flow Cytometry 1ul/1 million cells
Gene Accession No. P51449
Gene Alias MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR
Gene Symbols RORC
Host Species Mouse
Immunogen A portion of amino acids 1-50 of human ROR gamma was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 6097
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human, 86% in Mouse and Rat, 80% in Bovine, and 73% in Chicken.
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.