Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR gamma/RORC/NR1F3 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N224503J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N224503J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N224503J549 Supplier Novus Biologicals Supplier No. NBP224503JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ROR gamma/RORC/NR1F3 Polyclonal antibody specifically detects ROR gamma/RORC/NR1F3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen ROR gamma/RORC/NR1F3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR
Gene Symbols RORC
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human ROR gamma/RORC/NR1F3 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 6097.0
Test Specificity 100% homologous in human isoforms CRA_a).
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.