Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR gamma/RORC/NR1F3 Antibody (RORC/2942), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB392455 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392455 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392455 Supplier Novus Biologicals Supplier No. NBP308321AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROR gamma/RORC/NR1F3 Monoclonal antibody specifically detects ROR gamma/RORC/NR1F3 in Human samples. It is validated for ELISA, Protein Array

Specifications

Antigen ROR gamma/RORC/NR1F3
Applications ELISA, Protein Array
Classification Monoclonal
Clone RORC/2942
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias MGC129539, Nuclear receptor ROR gamma, Nuclear receptor ROR-gamma, Nuclear receptor RZR gamma, Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-Related Orphan Nuclear Receptor Variant 2, RAR-related orphan receptor C, RAR-related orphan receptor gamma, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma, Retinoid-related orphan receptor-gamma, ROR gamma, RORG, RORGMGC129539, RZRG, RZRGRZR-GAMMA, TOR
Host Species Mouse
Immunogen Recombinant full-length human ROR gamma/RORC/NR1F3 protein (Uniprot: P51449)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 6097
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.