Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR gamma t/RORC2/NR1F3 Antibody (1181A), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB347115 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347115 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347115 Supplier Novus Biologicals Supplier No. FAB61093MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ROR gamma t/RORC2/NR1F3 Monoclonal antibody specifically detects ROR gamma t/RORC2/NR1F3 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen ROR gamma t/RORC2/NR1F3
Applications Flow Cytometry
Classification Monoclonal
Clone 1181A
Conjugate mFluor Violet 450 SE
Dilution Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias IMD42;NR1F3;Nuclear receptor ROR-gamma;RORG;RZRG;RZR-GAMMA;TOR
Host Species Rabbit
Immunogen E.coli derived recombinant human ROR gamma T/RORC2/NR1F3, Met1-Gln100, Accession # P51449
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6097
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.