Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR1 Antibody (1011919), Biotin, Novus Biologicals™
SDP

Catalog No. NB366795 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366795 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366795 Supplier Novus Biologicals Supplier No. FAB20001B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROR1 Monoclonal specifically detects ROR1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen ROR1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 1011919
Conjugate Biotin
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias EC 2.7.10.1, MGC99659, neurotrophic tyrosine kinase receptor-related 1, Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1dJ537F10.1, receptor tyrosine kinase-like orphan receptor 1, tyrosine-protein kinase transmembrane receptor ROR1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ROR1, Ala24-Glu403, Accession # Q01973
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4919
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.