Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR1 Antibody (OTI3D11), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP273931A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273931A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273931A70 Supplier Novus Biologicals Supplier No. NBP273931AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROR1 Monoclonal specifically detects ROR1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen ROR1
Applications Western Blot
Classification Monoclonal
Clone OTI3D11
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 2.7.10.1, MGC99659, neurotrophic tyrosine kinase receptor-related 1, Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1dJ537F10.1, receptor tyrosine kinase-like orphan receptor 1, tyrosine-protein kinase transmembrane receptor ROR1
Gene Symbols ROR1
Host Species Mouse
Immunogen Full length recombinant protein of human ROR1 (NP_005003) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4919.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.