Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR2 Antibody (231512), PerCP, Novus Biologicals™
SDP

Catalog No. NB296882 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296882 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296882 Supplier Novus Biologicals Supplier No. FAB2064C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROR2 Monoclonal antibody specifically detects ROR2 in Human samples. It is validated for Western Blot

Specifications

Antigen ROR2
Applications Western Blot
Classification Monoclonal
Clone 231512
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ROR2, Val28-Gly403, Accession # AAA60276
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4920
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.