Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR2 Antibody (ROR2/1911), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB380026 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380026 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380026 Supplier Novus Biologicals Supplier No. NBP308532JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROR2 Monoclonal antibody specifically detects ROR2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen ROR2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ROR2/1911
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2
Host Species Mouse
Immunogen Human cultured keratinocytes (Uniprot: Q01974)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4920
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.