Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROR2 Rabbit anti-Mouse, Clone: 0B7X8, Novus Biologicals™
SDP

Catalog No. NB168220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168220 20 μg
NB168219 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168220 Supplier Novus Biologicals Supplier No. NBP31656420UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ROR2 Monoclonal antibody specifically detects ROR2 in Mouse samples. It is validated for Western Blot

Specifications

Antigen ROR2
Applications Western Blot
Classification Monoclonal
Clone 0B7X8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ROR2 (Q01974).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4920
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.