Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROS Antibody (4-6G), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309808 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309808 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309808 Supplier Novus Biologicals Supplier No. NBP250179MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROS Monoclonal antibody specifically detects ROS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional Assay, CyTOF

Specifications

Antigen ROS
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Functional Assay
Classification Monoclonal
Clone 4-6G
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias proto-oncogene tyrosine-protein kinase ROS, c-ros oncogene 1, receptor tyrosine kinase, c-ros-1, MCF3, ROS
Host Species Mouse
Immunogen Extracellular portion of ROS amino acid 1-285 fused to Fc, transiently expressed in 293 cells and purified using PtnA column chromatography.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6098
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.