Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROS Antibody (7-2B), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP250182G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250182G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250182G Supplier Novus Biologicals Supplier No. NBP250182G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROS Monoclonal specifically detects ROS in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen ROS
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 7-2B
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias proto-oncogene tyrosine-protein kinase ROS, c-ros oncogene 1 receptor tyrosine kinase, c-ros-1, MCF3, ROS
Gene Symbols ROS1
Host Species Mouse
Immunogen Extracellular portion of ROS amino acid 1-285 fused to Fc, transiently expressed in 293 cells and purified using PtnA column chromatography.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6098
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.