Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotavirus Antibody (531), Novus Biologicals™
SDP

Catalog No. NB10063039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063039 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063039 Supplier Novus Biologicals Supplier No. NB10063039

Mouse Monoclonal Antibody

Rotavirus Monoclonal specifically detects Rotavirus in Virus samples. It is validated for ELISA.

Specifications

Antigen Rotavirus
Applications ELISA
Classification Monoclonal
Clone 531
Conjugate Unconjugated
Dilution ELISA 1:20-1200
Formulation Phosphate buffered saline with 0.1% Sodium Azide
Gene Alias Major inner capsid protein VP6, VP6
Host Species Mouse
Immunogen Made to Viral ROTAVIRUS
Purification Method Protein A or G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-63039 detects intact Rotavirus strains RRV, Wa and Bovine. Rotavirus is a member of the Reoviridae family, and is the leading cause of diarrhoea hospitalisation among children worldwide. Seven major groups have been identified, of which three (groups A, B, and C) infect humans. Group A is the most common and widespread.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.