Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rotavirus Antibody, Novus Biologicals™
SDP

Catalog No. NB6001362 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001362 0.5 mL
NB018209 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6001362 Supplier Novus Biologicals Supplier No. NB6001362
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Rotavirus Polyclonal specifically detects Rotavirus in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Rotavirus
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 4.0 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation Phosphate Buffered Saline with 0.1% Sodium Azide
Gene Alias Major inner capsid protein VP6, VP6
Host Species Goat
Immunogen Bovine Nebraska Calf Diarrhea Virus.
Purification Method Protein G purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity This is specific against ICPs & late structural (virion) antigens. Uninfected cell reactivity is negative against HEp-2 cells and WI-38 cells by indirect immunofluorescence.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.