Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rotavirus Antibody, Novus Biologicals™

Goat Polyclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Rotavirus |
---|---|
Concentration | 4.0 mg/mL |
Dilution | Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Description
Rotavirus Polyclonal specifically detects Rotavirus in Virus samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Rotavirus | |
Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Virus | |
Major inner capsid protein VP6, VP6 | |
IgG | |
Protein G purified | |
This is specific against ICPs & late structural (virion) antigens. Uninfected cell reactivity is negative against HEp-2 cells and WI-38 cells by indirect immunofluorescence. |
4.0 mg/mL | |
Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Goat | |
Virology Bacteria and Parasites | |
Phosphate Buffered Saline with 0.1% Sodium Azide | |
Bovine Nebraska Calf Diarrhea Virus. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title