Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RP 001 hydrochloride
GREENER_CHOICE

Catalog No. 428910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-891-0 10 mg
42-895-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-891-0 Supplier Tocris Bioscience™ Supplier No. 4289/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent S1P1 agonist

Potent agonist of S1P1 receptors (EC50 = 9 pM). Induces internalization and polyubiquitination of S1P1 in vitro. Selective for S1P1 over S1P2 - S1P4.

Chemical Identifiers

CAS 1306761-53-4
Molecular Formula C24H25ClN4O4

Specifications

Quantity 10 mg
Formula Weight 468.93
Purity >98%
Molecular Weight (g/mol) 468.938
Chemical Name or Material RP 001 hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.