Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RP 67580
GREENER_CHOICE

Catalog No. p-4785455
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective NK1 antagonist

Potent and selective tachykinin NK1 receptor antagonist (Ki values are 2.9 nM and > 10 μM for rat NK1, and rat NK2 and NK3 receptors respectively). Displays higher affinity at rat and mouse than human receptors. Antinociceptive in vivo, possibly partly via inhibition of calcium channels.

Chemical Identifiers

CAS 135911-02-3
Molecular Formula C29H30N2O2

Specifications

Quantity 10 mg
Formula Weight 438.57
Purity >97%
Molecular Weight (g/mol) 438.571
Chemical Name or Material RP 67580
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.