Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RP1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222149SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222149SS 0.025 mL
NBP222149 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222149SS Supplier Novus Biologicals Supplier No. NBP222149SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RP1 Polyclonal specifically detects RP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen RP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.0 ug/ml, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:400
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P56715, P56716
Gene Alias DCDC4A, FLJ50293, FLJ55454, oxygen-regulated protein 1, retinitis pigmentosa 1 (autosomal dominant), retinitis pigmentosa 1 protein, retinitis pigmentosa RP1 protein
Gene Symbols RP1
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human RP1 protein (between residues 2000-2156) [UniProt P56715]
Molecular Weight of Antigen 240 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Hypoxia, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6101
Test Specificity Human and mouse. Predicted to react with monkey based upon 100% sequence homology.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.