Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RP1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB337558 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337558 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337558 Supplier Novus Biologicals Supplier No. NBP222149MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RP1 Polyclonal antibody specifically detects RP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen RP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DCDC4A, FLJ50293, FLJ55454, oxygen-regulated protein 1, retinitis pigmentosa 1 (autosomal dominant), retinitis pigmentosa 1 protein, retinitis pigmentosa RP1 protein
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human RP1 protein (between residues 2000-2156) [UniProt P56715]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hypoxia, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6101
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.