Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RP105/CD180 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB198336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB198336 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB198336 Supplier Novus Biologicals Supplier No. NBP176707V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RP105/CD180 Polyclonal antibody specifically detects RP105/CD180 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen RP105/CD180
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CD180 antigenMGC126233, CD180 molecule, LY64MGC126234, Ly78, Lymphocyte antigen 64, lymphocyte antigen-64, radioprotective, 105kDa, Radioprotective 105 kDa protein, RP105lymphocyte antigen 64 (mouse) homolog, radioprotective, 105kD
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the center of human RP105. The immunogen is located within amino acids 170 - 220 of RP105. Amino Acid Squence: HYISREDMRSLEQA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 4064
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.