Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RP105/CD180 Antibody (G28-8), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP262228A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP262228A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP262228A32 Supplier Novus Biologicals Supplier No. NBP262228AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RP105/CD180 Monoclonal specifically detects RP105/CD180 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen RP105/CD180
Applications Flow Cytometry
Classification Monoclonal
Clone G28-8
Conjugate Alexa Fluor 532
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD180 antigenMGC126233, CD180 molecule, LY64MGC126234, Ly78, Lymphocyte antigen 64, lymphocyte antigen-64, radioprotective, 105kDa, Radioprotective 105 kDa protein, RP105lymphocyte antigen 64 (mouse) homolog, radioprotective, 105kD
Gene Symbols CD180
Host Species Mouse
Immunogen Human tonsillar B cells
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4064
Test Specificity The antibody VI-01 reacts with vimentin, a 57 kDa intermediate filament expressed in variety of mesenchymal and mesodermal cell types. Cross-reactivity was found with smooth muscle desmin.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.