Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPA14 Antibody (14.2), Novus Biologicals™
SDP

Catalog No. NB100160 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100160 Supplier Novus Biologicals Supplier No. NB100160
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

RPA14 Monoclonal specifically detects RPA14 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen RPA14
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 14.2
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 2 ug/ml, Immunoprecipitation 0.1-2 ug/ml
Formulation PBS (pH 7.4) with No Preservative
Gene Alias REPA3Replication factor A protein 3, replication protein A 14 kDa subunit, replication protein A3 (14kD), replication protein A3, 14kDa, RF-A protein 3, RP-A p14, RPA14
Gene Symbols RPA3
Host Species Mouse
Immunogen Full-length human RPA-14 expressed in E. coli..
Molecular Weight of Antigen 14 kDa
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6119
Test Specificity NB100-160 is specific for human and mouse RPA14.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.